3-(Trifluormethyl)phenylhydraziniumchlorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Trifluormethyl)phenylhydrazin Hydrochlorid, 3-Hydrazinobenzotrifluorid Hydrochlorid, 3-(Trifluormethyl)phenylhydrazinhydrochlorid, 1-(3-(Trifluormethyl)phenyl)hydrazin Hydrochlorid

PC2707-10G PC2707-1G PC2707-5G
APOSPC2707-10GEA 23.2 EUR
APOSPC2707-10G APOSPC2707-1G APOSPC2707-5G
3-(Trifluormethyl)phenylhydraziniumchlorid 97%
3-(Trifluormethyl)phenylhydraziniumchlorid
Formel: C₇H₈ClF₃N₂
Molecular Weight: 212,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00100503
CAS-Nummer: 3107-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR