2-Chlor-5-fluorbenzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC2717-5G
PC2717-1G
PC2717-25G
APOSPC2717-5GEA
26.1
EUR
APOSPC2717-5G
APOSPC2717-1G
APOSPC2717-25G
2-Chlor-5-fluorbenzonitril 98%
2-Chlor-5-fluorbenzonitril
Formel:
C₇H₃ClFN Schmelzpunkt: 83…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094168 CAS-Nummer: 57381-56-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...