2-Fluor-4-iodbenzoesäure 95%

Lieferant: Apollo Scientific
Achtung

PC2740-1G PC2740-25G PC2740-5G
APOSPC2740-1GEA 21.9 EUR
APOSPC2740-1G APOSPC2740-25G APOSPC2740-5G
2-Fluor-4-iodbenzoesäure 95%
2-Fluoro-4-iodobenzoic acid
Formel: C₇H₄FIO₂
Molecular Weight: 266,0100 g/mol
Schmelzpunkt: 216…219 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094173
CAS-Nummer: 124700-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR