2,2'-Difluorbiphenyl

Lieferant: Apollo Scientific
Achtung

PC2748-1G PC2748-25G PC2748-5G
APOSPC2748-1GEA 87 EUR
APOSPC2748-1G APOSPC2748-25G APOSPC2748-5G
2,2'-Difluorbiphenyl
2,2'-Difluorbiphenyl
Formel: C₁₂H₈F₂
Molecular Weight: 190,19 g/mol
Schmelzpunkt: 117…121 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 388-82-9
UN: 3152
ADR: 9,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR