4-(Trifluormethoxy)zimtsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Trifluoromethoxy)cinnamic Acid
PC2756-1G
PC2756-500MG
APOSPC2756-1GEA
36.4
EUR
APOSPC2756-1G
APOSPC2756-500MG
4-(Trifluormethoxy)zimtsäure 96%
4-(Trifluormethoxy)zimtsäure
Formel:
C₁₀H₇F₃O₃ Molecular Weight: 232,1600 g/mol Schmelzpunkt: 185…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066338 CAS-Nummer: 783-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...