trans-2,5-Difluorzimtsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(2,5-Difluorphenyl)propensäure, 2,5-Difluorcinnamsäure

PC2762-25G PC2762-100G PC2762-5G
APOSPC2762-25GEA 77 EUR
APOSPC2762-25G APOSPC2762-100G APOSPC2762-5G
trans-2,5-Difluorzimtsäure 97%
trans-2,5-Difluorzimtsäure
Formel: C₉H₆F₂O₂
Molecular Weight: 184.14 g/mol
Schmelzpunkt: 138…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010318
CAS-Nummer: 112898-33-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR