2-Brom-4-fluor-6-nitrophenol 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC2773-1G
PC2773-5G
PC2773-25G
APOSPC2773-1GEA
21.9
EUR
APOSPC2773-1G
APOSPC2773-5G
APOSPC2773-25G
2-Brom-4-fluor-6-nitrophenol 95%
2-Brom-4-fluor-6-nitrophenol
Formel:
C₆H₃BrFNO₃ Molecular Weight: 235,9953 g/mol Schmelzpunkt: 67 °C Flammpunkt: 98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094182 CAS-Nummer: 320-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...