2-Amino-4,5-difluorphenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC2777-1G
PC2777-250MG
APOSPC2777-1GEA
181
EUR
APOSPC2777-1G
APOSPC2777-250MG
2-Amino-4,5-difluorphenol 98%
2-Amino-4,5-difluorphenol
Formel:
C₆H₅F₂NO Molecular Weight: 145,11 g/mol Schmelzpunkt: 135…140 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03094185 CAS-Nummer: 163734-01-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...