Fmoc-Lys(Tfa)-OH
Lieferant: Apollo Scientific
Synonyme:
Nα-Fmoc-Nepsilon-trifluoracetyl-L-lysine
PC27880-25G
APOSPC27880-25GEA
97.3
EUR
APOSPC27880-25G
Fmoc-Lys(Tfa)-OH
Fmoc-Lys(Tfa)-OH
Formel:
C₂₃H₂₃F₃N₂O₅ Molecular Weight: 464,441 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153360 CAS-Nummer: 76265-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...