(2,4-Difluorphenoxy)acetonitril 95%
Lieferant: Apollo Scientific
PC28200-1G
PC28200-5G
APOSPC28200-1GEA
270
EUR
APOSPC28200-1G
APOSPC28200-5G
(2,4-Difluorphenoxy)acetonitril 95%
(2,4-Difluorphenoxy)acetonitril
Formel:
C₈H₅F₂NO Lagertemperatur: Raumtemperatur |
CAS-Nummer:
942473-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...