2-Methoxy-4-nitro-1-(trifluoromethyl)benzene 95%

Lieferant: Apollo Scientific
Gefahr

PC28201-1G PC28201-250MG PC28201-5G
APOSPC28201-1GEA 190 EUR
APOSPC28201-1G APOSPC28201-250MG APOSPC28201-5G
2-Methoxy-4-nitro-1-(trifluoromethyl)benzene 95%
2-Methoxy-4-nitro-1-(trifluoromethyl)benzene
Formel: C₈H₆F₃NO₃
Molecular Weight: 221.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112485
CAS-Nummer: 453560-74-2
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR