Methyl-5-amino-2-fluorobenzenecarboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 5-amino-2-fluorobenzoate

PC28215-25G PC28215-5G
APOSPC28215-25GEA 417 EUR
APOSPC28215-25G APOSPC28215-5G
Methyl-5-amino-2-fluorobenzenecarboxylate 95%
Methyl-5-amino-2-fluorobenzenecarboxylate
Formel: C₈H₈FNO₂
Molecular Weight: 169,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037918
CAS-Nummer: 56741-34-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR