3-Fluoro-6-methoxypyridine-2-carboxylic acid 98%
Lieferant: Apollo Scientific
PC28219-1G
PC28219-5G
APOSPC28219-1GEA
445
EUR
APOSPC28219-1G
APOSPC28219-5G
3-Fluoro-6-methoxypyridine-2-carboxylic acid 98%
3-Fluoro-6-methoxypyridine-2-carboxylic acid
Formel:
C₇H₆FNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185873 CAS-Nummer: 1214328-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...