4-Ethyl-5-fluoro-6-hydroxypyrimidine

Lieferant: Apollo Scientific
Achtung

PC2822-1G PC2822-25G PC2822-5G
APOSPC2822-1GEA 21.9 EUR
APOSPC2822-1G APOSPC2822-25G APOSPC2822-5G
4-Ethyl-5-fluoro-6-hydroxypyrimidine
4-Ethyl-5-fluoro-6-hydroxypyrimidine
Formel: C₆H₇FN₂O
Molecular Weight: 142.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08063343
CAS-Nummer: 137234-87-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR