2-{[(4-Fluorophenyl)methyl]amino}-2-methylpropanenitrile 95%

Lieferant: Apollo Scientific

PC28223-1G PC28223-5G
APOSPC28223-1GEA 180 EUR
APOSPC28223-1G APOSPC28223-5G
2-{[(4-Fluorophenyl)methyl]amino}-2-methylpropanenitrile 95%
2-{[(4-Fluorophenyl)methyl]amino}-2-methylpropanenitrile
Formel: C₁₁H₁₃FN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10014477
CAS-Nummer: 1018539-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR