Ethyl-4-cyano-3-fluorobenzenecarboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 4-cyano-3-fluorobenzoate
PC28230-1G
PC28230-5G
APOSPC28230-1GEA
78.3
EUR
APOSPC28230-1G
APOSPC28230-5G
Ethyl-4-cyano-3-fluorobenzenecarboxylate 95%
Ethyl-4-cyano-3-fluorobenzenecarboxylate
Formel:
C₁₀H₈FNO₂ Molecular Weight: 193,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09607944 CAS-Nummer: 952183-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...