2-Amino-5-fluorbenzolsulfonamid

Lieferant: Apollo Scientific
Achtung

PC2824-1G PC2824-5G
APOSPC2824-1GEA 149 EUR
APOSPC2824-1G APOSPC2824-5G
2-Amino-5-fluorbenzolsulfonamid
2-Amino-5-fluorbenzolsulfonamid
Formel: C₆H₇FN₂O₂S
Molecular Weight: 190,200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08741368
CAS-Nummer: 1992-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR