5-Fluor-2-pyridincarbonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Cyan-5-fluorpyridin, 5-Fluor-2-pyridincarbonsäurenitril
PC2830-5G
PC2830-100G
PC2830-25G
APOSPC2830-5GEA
33.5
EUR
APOSPC2830-5G
APOSPC2830-100G
APOSPC2830-25G
5-Fluor-2-pyridincarbonitril 97%
5-Fluor-2-pyridincarbonitril
Formel:
C₆H₃FN₂ Molecular Weight: 122,1 g/mol Schmelzpunkt: 35…41 °C Flammpunkt: 98,9 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08741371 CAS-Nummer: 327056-62-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...