5-Fluor-1-naphthoesäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC2868-1G
PC2868-5G
APOSPC2868-1GEA
132
EUR
APOSPC2868-1G
APOSPC2868-5G
5-Fluor-1-naphthoesäure 99%
5-Fluor-1-naphthoesäure
Formel:
C₁₁H₇FO₂ Schmelzpunkt: 213…216 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
573-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...