5-Fluor-1-naphthaldehyd 98%

Lieferant: Apollo Scientific
Achtung

PC2874-1G PC2874-5G
APOSPC2874-1GEA 132 EUR
APOSPC2874-1G APOSPC2874-5G
5-Fluor-1-naphthaldehyd 98%
5-Fluor-1-naphthaldehyd
Formel: C₁₁H₇FO
Lagertemperatur: Raumtemperatur
MDL: MFCD09258965
CAS-Nummer: 110931-86-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR