(2,3-Difluorphenyl)essigsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2,3-Difluorophenyl)acetic acid

PC2874A-100G PC2874A-25G PC2874A-5G
APOSPC2874A-100GEA 132 EUR
APOSPC2874A-100G APOSPC2874A-25G APOSPC2874A-5G
(2,3-Difluorphenyl)essigsäure 97%
(2,3-Difluorphenyl)essigsäure
Formel: (C₆H₃F₂)CH₂COOH
Molecular Weight: 172,13 g/mol
Schmelzpunkt: 116…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040968
CAS-Nummer: 145689-41-4
EINECS: 279-709-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR