(2,4-Difluorphenyl)essigsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4-Difluorophenylacetic acid
PC2874B-1G
PC2874B-25G
PC2874B-5G
APOSPC2874B-1GEA
14.7
EUR
APOSPC2874B-1G
APOSPC2874B-25G
APOSPC2874B-5G
(2,4-Difluorphenyl)essigsäure 97%
(2,4-Difluorphenyl)essigsäure
Formel:
(C₆H₃F₂)CH₂COOH Molecular Weight: 172,13 g/mol Schmelzpunkt: 115…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009999 CAS-Nummer: 81228-09-3 EINECS: 279-709-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...