(3,5-Difluorphenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Difluorophenylacetic acid

PC2874EC-100G PC2874EC-25G PC2874EC-5G
APOSPC2874EC-100GEA 183 EUR
APOSPC2874EC-100G APOSPC2874EC-25G APOSPC2874EC-5G
(3,5-Difluorphenyl)essigsäure 98%
(3,5-Difluorphenyl)essigsäure
Formel: (C₆H₃F₂)CH₂COOH
Molecular Weight: 172,13 g/mol
Schmelzpunkt: 68…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010316
CAS-Nummer: 105184-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR