(3,5-Difluorphenyl)acetonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Difluorbenzylcyanid

PC2874EL-100G PC2874EL-10G PC2874EL-25G
APOSPC2874EL-100GEA 184 EUR
APOSPC2874EL-100G APOSPC2874EL-10G APOSPC2874EL-25G
(3,5-Difluorphenyl)acetonitril 98%
(3,5-Difluorphenyl)acetonitril
Formel: C₈H₅F₂N
Molecular Weight: 153,13 g/mol
Schmelzpunkt: 34…36 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061278
CAS-Nummer: 122376-76-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR