3,5-Difluor-DL-phenylalanin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2874L-1G
PC2874L-250MG
APOSPC2874L-1GEA
29
EUR
APOSPC2874L-1G
APOSPC2874L-250MG
3,5-Difluor-DL-phenylalanin 97%
3,5-Difluor-DL-phenylalanin
Formel:
C₉H₉F₂NO₂ Molecular Weight: 201,1701 g/mol Schmelzpunkt: 243…245 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061308 CAS-Nummer: 32133-37-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...