N-(2,4-Difluorphenyl)maleamidsäure 97%
Lieferant: Apollo Scientific
PC2877-25G
PC2877-5G
APOSPC2877-25GEA
669
EUR
APOSPC2877-25G
APOSPC2877-5G
N-(2,4-Difluorphenyl)maleamidsäure 97%
N-(2,4-Difluorphenyl)maleamidsäure
Formel:
C₁₀H₇F₂NO₃ Schmelzpunkt: 195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082911 CAS-Nummer: 6954-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...