2',5'-Difluorpropiophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2,5-Difluorophenyl)propan-1-one

PC2881-10G PC2881-1G PC2881-5G
APOSPC2881-10GEA 50.9 EUR
APOSPC2881-10G APOSPC2881-1G APOSPC2881-5G
2',5'-Difluorpropiophenon 98%
2',5'-Difluorpropiophenon
Formel: C₉H₈F₂O
Molecular Weight: 170,16 g/mol
Siedepunkt: 39…40 °C/0,5mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00015509
CAS-Nummer: 29112-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR