2',6'-Difluorpropiophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2,6-Difluorophenyl)propan-1-one
PC2882-10G
PC2882-1G
PC2882-5G
APOSPC2882-10GEA
69.6
EUR
APOSPC2882-10G
APOSPC2882-1G
APOSPC2882-5G
2',6'-Difluorpropiophenon
2',6'-Difluorpropiophenon
Formel:
C₉H₈F₂O Molecular Weight: 170.16 g/mol Siedepunkt: 70…71 °C/4mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015506 CAS-Nummer: 85068-31-1 EINECS: 285-293-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...