3-Fluor-4-methoxyphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Fluoro-4-methoxyphenyl)boronic acid, 3-Fluoro-4-methoxybenzeneboronic acid

PC2902-25G PC2902-5G PC2902-1G
APOSPC2902-25GEA 51.1 EUR
APOSPC2902-25G APOSPC2902-5G APOSPC2902-1G
3-Fluor-4-methoxyphenylboronsäure 97%
3-Fluor-4-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
Molecular Weight: 169.95 g/mol
Schmelzpunkt: 211…213 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00807404
CAS-Nummer: 149507-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR