3-Chlor-5-fluorbenzonitril
Lieferant: Apollo Scientific
|
Achtung
|
PC2904-100G
PC2904-25G
PC2904-5G
APOSPC2904-100GEA
404
EUR
APOSPC2904-100G
APOSPC2904-25G
APOSPC2904-5G
3-Chlor-5-fluorbenzonitril
3-Chlor-5-fluorbenzonitril
Formel:
C₇H₃ClFN Molecular Weight: 155,56 g/mol Schmelzpunkt: 54…59 °C Flammpunkt: 89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407967 CAS-Nummer: 327056-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...