5-Brom-2-fluoranisol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-1-fluoro-2-methoxybenzene
PC2913-25G
PC2913-5G
APOSPC2913-25GEA
33.5
EUR
APOSPC2913-25G
APOSPC2913-5G
5-Brom-2-fluoranisol 98%
5-Brom-2-fluoranisol
Formel:
C₇H₆BrFO Molecular Weight: 205.03 g/mol Siedepunkt: 82…85 °C Flammpunkt: 98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861130 CAS-Nummer: 103291-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...