4-Brom-2-chlorbenzotrifluorid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC2915-25G
PC2915-5G
PC2915-100G
APOSPC2915-25GEA
33.5
EUR
APOSPC2915-25G
APOSPC2915-5G
APOSPC2915-100G
4-Brom-2-chlorbenzotrifluorid 98%
4-Brom-2-chlorbenzotrifluorid
Formel:
C₇H₃BrClF₃ Molecular Weight: 259,45 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08459292 CAS-Nummer: 467435-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...