3-Chlor-5-(trifluormethoxy)benzoesäure 98%

Lieferant: Apollo Scientific

PC2933-10G PC2933-5G PC2933-1G PC2933-25G
APOSPC2933-10GEA 314 EUR
APOSPC2933-10G APOSPC2933-5G APOSPC2933-1G APOSPC2933-25G
3-Chlor-5-(trifluormethoxy)benzoesäure 98%
3-Chloro-5-(trifluoromethoxy)benzoic acid
Formel: C₈H₄ClF₃O₃
Molecular Weight: 240,57 g/mol
Schmelzpunkt: 65…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06660306
CAS-Nummer: 433926-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR