2-Amino-4-bromo-6-(trifluoromethyl)phenylamine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-3-(trifluoromethyl)benzene-1,2-diamine, 5-Bromo-2,3-diaminobenzotrifluoride
PC2934-1G
PC2934-500MG
PC2934-5G
APOSPC2934-1GEA
52.2
EUR
APOSPC2934-1G
APOSPC2934-500MG
APOSPC2934-5G
2-Amino-4-bromo-6-(trifluoromethyl)phenylamine 97%
2-Amino-4-bromo-6-(trifluoromethyl)phenylamine
Formel:
C₇H₆BrF₃N₂ Molecular Weight: 255,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08741393 CAS-Nummer: 157026-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...