2-(Difluoromethoxy)-6-fluoropyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC2974-1G
PC2974-5G
PC2974-25G
APOSPC2974-1GEA
134
EUR
APOSPC2974-1G
APOSPC2974-5G
APOSPC2974-25G
2-(Difluoromethoxy)-6-fluoropyridine 95%
2-(Difluoromethoxy)-6-fluoropyridine
Formel:
C₆H₄F₃NO Lagertemperatur: Kühlschrank |
MDL:
MFCD09258966 CAS-Nummer: 947534-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...