4-(Trifluormethyl)benzolsulfonamid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
α,α,α-Trifluor-p-toluolsulfonamid, 4-(Trifluormethyl)benzolsulfonsäureamid
PC2981-25G
PC2981-5G
APOSPC2981-25GEA
65.4
EUR
APOSPC2981-25G
APOSPC2981-5G
4-(Trifluormethyl)benzolsulfonamid
4-(Trifluormethyl)benzolsulfonamid
Formel:
C₇H₆F₃NO₂S Molecular Weight: 225,19 g/mol Schmelzpunkt: 181 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159251 CAS-Nummer: 830-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...