3-Chloro-4-fluoropyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC2990-1G
PC2990-5G
APOSPC2990-1GEA
181
EUR
APOSPC2990-1G
APOSPC2990-5G
3-Chloro-4-fluoropyridine
3-Chloro-4-fluoropyridine
Formel:
C₅H₃ClFN Molecular Weight: 131.54 g/mol Schmelzpunkt: 16 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04973418 CAS-Nummer: 883107-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...