2-Carboxy-5-fluorphenylboronsäure

Lieferant: Apollo Scientific

Synonyme: 2-Carboxy-5-flourophenylboronic acid

PC2996-1G PC2996-5G
APOSPC2996-1GEA 87.3 EUR
APOSPC2996-1G APOSPC2996-5G
2-Carboxy-5-fluorphenylboronsäure
2-Carboxy-5-fluorphenylboronsäure
Formel: C₇H₆BFO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD06656266
CAS-Nummer: 874290-62-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR