3-(N-(4-(Trifluoromethoxy)phenyl)carbamoyl)prop-2-enoic acid

Lieferant: Apollo Scientific

Synonyme: 4-Oxo-4-{[4-(trifluoromethoxy)phenyl]amino}but-2-enoic acid

PC300522-1G PC300522-5G
APOSPC300522-1GEA 94.4 EUR
APOSPC300522-1G APOSPC300522-5G
3-(N-(4-(Trifluoromethoxy)phenyl)carbamoyl)prop-2-enoic acid
3-(N-(4-(Trifluoromethoxy)phenyl)carbamoyl)prop-2-enoic acid
Formel: C₁₁H₈F₃NO₄
Molecular Weight: 275,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00169478
CAS-Nummer: 298217-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR