Ethyl-6-(trifluoromethyl)-1-benzothiophene-2-carboxylate

Lieferant: Apollo Scientific

PC300545-1G PC300545-5G PC300545-10G
APOSPC300545-1GEA 154 EUR
APOSPC300545-1G APOSPC300545-5G APOSPC300545-10G
Ethyl-6-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Ethyl-6-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Formel: C₁₂H₉F₃O₂S
Molecular Weight: 274,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100110
CAS-Nummer: 1132650-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR