Methyl-3-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanoate

Lieferant: Apollo Scientific

PC300548-1G PC300548-5G
APOSPC300548-1GEA 109 EUR
APOSPC300548-1G APOSPC300548-5G
Methyl-3-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanoate
Methyl-3-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanoate
Formel: C₁₀H₉ClF₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD03153507
CAS-Nummer: 380453-90-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR