Ethyl-3-amino-5-(trifluoromethyl)-1-benzothiophene-2-carboxylate

Lieferant: Apollo Scientific

PC300550-1G PC300550-5G
APOSPC300550-1GEA 0 EUR
APOSPC300550-1G APOSPC300550-5G
Ethyl-3-amino-5-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Ethyl-3-amino-5-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Formel: C₁₂H₁₀F₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11505029
CAS-Nummer: 1217018-83-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR