3-Chloro-N-ethyl-5-(trifluoromethyl)-2-pyridinamine
Lieferant: Apollo Scientific
|
Achtung
|
PC300561-1G
PC300561-5G
APOSPC300561-1GEA
58
EUR
APOSPC300561-1G
APOSPC300561-5G
3-Chloro-N-ethyl-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-ethyl-5-(trifluoromethyl)-2-pyridinamine
Formel:
C₈H₈ClF₃N₂ Molecular Weight: 224,61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11122453 CAS-Nummer: 1042499-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...