(4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}phenyl)methanol

Lieferant: Apollo Scientific

PC300595-1G PC300595-5G
APOSPC300595-1GEA 149 EUR
APOSPC300595-1G APOSPC300595-5G
(4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}phenyl)methanol
(4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}phenyl)methanol
Formel: C₁₃H₁₀F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10703545
CAS-Nummer: 1086376-57-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR