3-Amino-2-methyl-6-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methyl-6-(trifluoromethyl)pyridin-3-amine

PC300644-1G PC300644-250MG
APOSPC300644-1GEA 116 EUR
APOSPC300644-1G APOSPC300644-250MG
3-Amino-2-methyl-6-(trifluoromethyl)pyridine
3-Amino-2-methyl-6-(trifluoromethyl)pyridine
Formel: C₇H₇F₃N₂
Molecular Weight: 176,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01311991
CAS-Nummer: 383907-17-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR