2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1-piperazinyl}-1-ethanol

Lieferant: Apollo Scientific

PC300675-1G PC300675-5G PC300675-10G
APOSPC300675-1GEA 149 EUR
APOSPC300675-1G APOSPC300675-5G APOSPC300675-10G
2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1-piperazinyl}-1-ethanol
2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1-piperazinyl}-1-ethanol
Formel: C₁₂H₁₅ClF₃N₃O
Molecular Weight: 309.72 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00203913
CAS-Nummer: 215434-40-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR