2-Fluor-4-(trifluormethyl)phenylboronsäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-4-(trifluoromethyl)benzeneboronic acid
PC3007-25G
PC3007-5G
PC3007-1G
APOSPC3007-25GEA
89.9
EUR
APOSPC3007-25G
APOSPC3007-5G
APOSPC3007-1G
2-Fluor-4-(trifluormethyl)phenylboronsäure 99%
2-Fluor-4-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BF₄O₂ Molecular Weight: 207,92 g/mol Schmelzpunkt: 113…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094995 CAS-Nummer: 503309-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...