N-Butyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine

Lieferant: Apollo Scientific

Synonyme: 2-(Butylamino)-3-chloro-5-(trifluoromethyl)pyridine

PC300792-1G PC300792-250MG PC300792-5G
APOSPC300792-1GEA 231 EUR
APOSPC300792-1G APOSPC300792-250MG APOSPC300792-5G
N-Butyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine
N-Butyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine
Formel: C₁₀H₁₂ClF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11119963
CAS-Nummer: 1041535-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR