2-(Dimethylamino)-6-fluorbenzonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC3009J-1G
APOSPC3009J-1GEA
109
EUR
APOSPC3009J-1G
2-(Dimethylamino)-6-fluorbenzonitril 97%
2-(Dimethylamino)-6-fluorbenzonitril
Formel:
C₉H₉FN₂ Siedepunkt: 131…135 °C/5mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052221 CAS-Nummer: 96994-73-9 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...