3-(4-Fluorophenyl)propanal

Lieferant: Apollo Scientific

PC301205-1G PC301205-5G
APOSPC301205-1GEA 292 EUR
APOSPC301205-1G APOSPC301205-5G
3-(4-Fluorophenyl)propanal
3-(4-Fluorophenyl)propanal
Formel: C₉H₉FO
Molecular Weight: 152.1656 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08064186
CAS-Nummer: 63416-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR